Learn More
MSC Peerless Chain Welded Transport Chain, 5/16 in, Grade 70, Carbon Steel, Yellow Zinc
Catalog No. 5000532110
Encompass
The Peerless Chain Welded Transport Chain is a durable Grade 70 chain, constructed from carbon steel with a yellow zinc finish for corrosion resistance. It features a 5/16 inch trade size and is supplied in a 550 ft coil, priced by the foot.
  • Trade size 5/16 in
  • Transport chain link type
  • Made from carbon steel
  • Yellow zinc finish for corrosion resistance
  • Grade 70 for high strength
  • Overall length 550 ft
  • Capacity of 4,700 lb
  • Inside width 0.47 in
  • Inside length 1.10 in
  • Available as cut-to-length items
Catalog No. 50-005-32110 Supplier MSC Supplier No. 84462928
The Peerless Chain Welded Transport Chain is a durable Grade 70 chain, constructed from carbon steel with a yellow zinc finish for corrosion resistance. It features a 5/16 inch trade size and is supplied in a 550 ft coil, priced by the foot.
  • Trade size 5/16 in
  • Transport chain link type
  • Made from carbon steel
  • Yellow zinc finish for corrosion resistance
  • Grade 70 for high strength
  • Overall length 550 ft
  • Capacity of 4,700 lb
  • Inside width 0.47 in
  • Inside length 1.10 in
  • Available as cut-to-length items