Learn More
Biolegend PE/Fire™ 810 anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 329755
Encompass_Preferred
PE/Fire™ 810 anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-213-1915