Learn More
Crescent Chemical Co Inc PEFABLOC SC
SDP
Supplier:  Crescent Chemical Co Inc SE31682.01
Encompass_Preferred
PEFABLOC SC Protease inhibitor with major advantages over other inhibitors: Excellent stability at neutral pH. Ready solubility in aqueous buffers; LD50 (oral) 2800 mg/kg. Broad specificity for serine proteases. Minimum effect on cell growth. Size - 100
Catalog No. 50-247-985