Learn More
Crescent Chemical Co Inc PEFABLOC SC
SDP
Catalog No. 50247986
Encompass_Preferred
PEFABLOC SC Protease inhibitor with major advantages over other inhibitors: Excellent stability at neutral pH. Ready solubility in aqueous buffers; LD50 (oral) 2800 mg/kg. Broad specificity for serine proteases. Minimum effect on cell growth. Size - 500
Catalog No. 50-247-986 Supplier Crescent Chemical Co Inc Supplier No. SE31682.02
Add to Cart
Add to Cart
PEFABLOC SC Protease inhibitor with major advantages over other inhibitors: Excellent stability at neutral pH. Ready solubility in aqueous buffers; LD50 (oral) 2800 mg/kg. Broad specificity for serine proteases. Minimum effect on cell growth. Size - 500