Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pefabloc™ SC, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013882 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3882 100 mg
11-101-3883 500 mg
11-101-3881 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-3882 Supplier MilliporeSigma Supelco Supplier No. 76307100MG
Add to Cart
Add to Cart

Pefabloc(R) SC is a trypsin inhibitor, which can be used in the synthesis of its arginine-like analogue, 4-(2-guanidinoethyl)benzenesulfonyl fluoride.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Product Line Pefabloc
CAS 30827-99-7
Physical Form Powder/Crystals
Linear Formula C8H10 FNO2 S · HCl
MDL Number MFCD00132962
Synonym 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride; AEBSF
Type Analytical Standard
Quantity 100 mg
Formula Weight 239.69
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less

Pefabloc is a registered trademark of Pentapharm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.