Learn More
Medchemexpress LLC Pefloxacin-d3 | 2733455-58-6 | 99.4% | 336.38 g/mol | C17H17D3FN3O3 | 10 MG
SDP
Catalog No. 5000296823
Encompass_Preferred
Pefloxacin-d3 is the deuterium-labeled analogue of the fluoroquinolone antibiotic pefloxacin. It inhibits DNA gyrase and interferes with topoisomerase I-mediated DNA relaxation, and is supplied as a high-purity stable isotope for analytical and research use.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity to support analytical accuracy.
  • Suitable as a tracer in infection and pharmacokinetic studies.
  • Provided as a laboratory-grade powder for analytical workflows.
Catalog No. 50-002-96823 Supplier Medchemexpress LLC Supplier No. HYB0147S110MG
Pefloxacin-d3 is the deuterium-labeled analogue of the fluoroquinolone antibiotic pefloxacin. It inhibits DNA gyrase and interferes with topoisomerase I-mediated DNA relaxation, and is supplied as a high-purity stable isotope for analytical and research use.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity to support analytical accuracy.
  • Suitable as a tracer in infection and pharmacokinetic studies.
  • Provided as a laboratory-grade powder for analytical workflows.