Learn More
Medchemexpress LLC Pefloxacin-d3 | 2733455-58-6 | 99.4% | 5 MG
SDP
Catalog No. 5000450701
Encompass_Preferred
Pefloxacin-d3 (Pefloxacinium-d3) is a deuterium-labeled version of the broad-spectrum antibiotic Pefloxacin. It blocks DNA replication by inhibiting DNA gyrase and also inhibits DNA relaxation catalyzed by topoisomerase I with an IC50 of 45 μg/mL. This compound demonstrates antibacterial activity against various bacteria such as *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*.
  • Deuterium labeled version of Pefloxacin
  • Broad-spectrum antibiotic
  • Inhibits DNA gyrase, blocking DNA replication
  • Inhibits DNA relaxation catalyzed by topoisomerase I
  • Exhibits antibacterial activity against *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*
  • Shows anti-*Plasmodium yoelii* infection activity
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50701 Supplier Medchemexpress LLC Supplier No. HYB0147S15MG
Pefloxacin-d3 (Pefloxacinium-d3) is a deuterium-labeled version of the broad-spectrum antibiotic Pefloxacin. It blocks DNA replication by inhibiting DNA gyrase and also inhibits DNA relaxation catalyzed by topoisomerase I with an IC50 of 45 μg/mL. This compound demonstrates antibacterial activity against various bacteria such as *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*.
  • Deuterium labeled version of Pefloxacin
  • Broad-spectrum antibiotic
  • Inhibits DNA gyrase, blocking DNA replication
  • Inhibits DNA relaxation catalyzed by topoisomerase I
  • Exhibits antibacterial activity against *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*
  • Shows anti-*Plasmodium yoelii* infection activity
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS