Learn More
Apexbio Technology LLC Pefloxacin Mesylate 70458-95-6 100mg
SDP
Catalog No. 502240540
Encompass_Preferred
Pefloxacin Mesylate (CAS 70458-95-6) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) It is designed to inhibit bacterial DNA gyrase activity thereby interfering with DNA replication and transcription Pefloxacin Mesylate exerts its biological activity primarily through inhibition of DNA gyrase In biochemical research assays Pefloxacin Mesylate demonstrates inhibitory activity with IC50 values in vitro in the low micromolar range depending on bacterial species and assay conditions Based on these pharmacological properties Pefloxacin Mesylate holds research potential in microbiological assays antimicrobial susceptibility tests and studies of bacterial enzyme inhibition
This item is not returnable. View return policy
Catalog No. 50-224-0540 Supplier Apexbio Technology LLC Supplier No. B2003100
Pefloxacin Mesylate (CAS 70458-95-6) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) It is designed to inhibit bacterial DNA gyrase activity thereby interfering with DNA replication and transcription Pefloxacin Mesylate exerts its biological activity primarily through inhibition of DNA gyrase In biochemical research assays Pefloxacin Mesylate demonstrates inhibitory activity with IC50 values in vitro in the low micromolar range depending on bacterial species and assay conditions Based on these pharmacological properties Pefloxacin Mesylate holds research potential in microbiological assays antimicrobial susceptibility tests and studies of bacterial enzyme inhibition