Learn More
Apexbio Technology LLC Pefloxacin Mesylate 70458-95-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093057
Encompass_Preferred
Pefloxacin Mesylate (CAS 70458-95-6) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) It is designed to inhibit bacterial DNA gyrase activity thereby interfering with DNA replication and transcription Pefloxacin Mesylate exerts its biological activity primarily through inhibition of DNA gyrase In biochemical research assays Pefloxacin Mesylate demonstrates inhibitory activity with IC50 values in vitro in the low micromolar range depending on bacterial species and assay conditions Based on these pharmacological properties Pefloxacin Mesylate holds research potential in microbiological assays antimicrobial susceptibility tests and studies of bacterial enzyme inhibition
Catalog No. 50-000-93057 Supplier Apexbio Technology LLC Supplier No. B20035.1
Pefloxacin Mesylate (CAS 70458-95-6) is a small-molecule inhibitor targeting bacterial DNA gyrase (topoisomerase II) It is designed to inhibit bacterial DNA gyrase activity thereby interfering with DNA replication and transcription Pefloxacin Mesylate exerts its biological activity primarily through inhibition of DNA gyrase In biochemical research assays Pefloxacin Mesylate demonstrates inhibitory activity with IC50 values in vitro in the low micromolar range depending on bacterial species and assay conditions Based on these pharmacological properties Pefloxacin Mesylate holds research potential in microbiological assays antimicrobial susceptibility tests and studies of bacterial enzyme inhibition