Learn More
Medchemexpress LLC Pefloxacin mesylate | 70458-95-6 | 99.7% | 1 G
SDP
Catalog No. 5000450699
Encompass_Preferred
Pefloxacin mesylate is a broad-spectrum antibiotic that blocks DNA replication by inhibiting DNA gyrase and DNA relaxation catalyzed by topoisomerase I. It exhibits antibacterial activity against various bacteria such as *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*. It has also shown activity against *Plasmodium yoelii* infection and can increase UVA-induced edema and immunosuppression, making it suitable for infection studies.
  • Inhibits DNA replication by blocking DNA gyrase
  • Inhibits DNA relaxation catalyzed by topoisomerase I (IC50 of 45 μg/mL)
  • Exhibits broad-spectrum antibacterial activity
  • Active against *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*
  • Demonstrates activity against *Plasmodium yoelii* infection
  • Increases UVA-induced edema and immunosuppression
  • Suitable for infection studies
Catalog No. 50-004-50699 Supplier Medchemexpress LLC Supplier No. HYB0147A1G
Add to Cart
Add to Cart
Pefloxacin mesylate is a broad-spectrum antibiotic that blocks DNA replication by inhibiting DNA gyrase and DNA relaxation catalyzed by topoisomerase I. It exhibits antibacterial activity against various bacteria such as *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*. It has also shown activity against *Plasmodium yoelii* infection and can increase UVA-induced edema and immunosuppression, making it suitable for infection studies.
  • Inhibits DNA replication by blocking DNA gyrase
  • Inhibits DNA relaxation catalyzed by topoisomerase I (IC50 of 45 μg/mL)
  • Exhibits broad-spectrum antibacterial activity
  • Active against *Escherichia coli*, *Pseudomonas aeruginosa*, and *Bacteroides fragilis*
  • Demonstrates activity against *Plasmodium yoelii* infection
  • Increases UVA-induced edema and immunosuppression
  • Suitable for infection studies