Learn More
Medchemexpress LLC Pefloxacin mesylate | 70458-95-6 | 99.7% | 5 G
SDP
Catalog No. 5000450700
Encompass_Preferred
Pefloxacin mesylate is a broad-spectrum antibiotic that inhibits DNA replication by blocking DNA gyrase and DNA relaxation by topoisomerase I (IC50 of 45 μg/mL). It is effective against Escherichia coli, Pseudomonas aeruginosa, and Bacteroides fragilis. This compound also shows anti-Plasmodium yoelii infection activity and is suitable for infection studies.
  • Broad-spectrum antibiotic.
  • Inhibits DNA gyrase and topoisomerase I.
  • Active against Escherichia coli, Pseudomonas aeruginosa, and Bacteroides fragilis.
  • Effective against Plasmodium yoelii infections.
  • Reduces bacterial load in gram-negative bacilli infections.
  • Increases survival rate in Plasmodium yoelii-infected mice.
Catalog No. 50-004-50700 Supplier Medchemexpress LLC Supplier No. HYB0147A5G
Pefloxacin mesylate is a broad-spectrum antibiotic that inhibits DNA replication by blocking DNA gyrase and DNA relaxation by topoisomerase I (IC50 of 45 μg/mL). It is effective against Escherichia coli, Pseudomonas aeruginosa, and Bacteroides fragilis. This compound also shows anti-Plasmodium yoelii infection activity and is suitable for infection studies.
  • Broad-spectrum antibiotic.
  • Inhibits DNA gyrase and topoisomerase I.
  • Active against Escherichia coli, Pseudomonas aeruginosa, and Bacteroides fragilis.
  • Effective against Plasmodium yoelii infections.
  • Reduces bacterial load in gram-negative bacilli infections.
  • Increases survival rate in Plasmodium yoelii-infected mice.