Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pefloxacin Mesylate Dihydrate, G-Biosciences
SCI_ED

Catalog No. S157231
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S157231 10 g
S139745 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S157231 Supplier G-Biosciences Supplier No. RC1084
Add to Cart
Add to Cart

Perfloxacin is a fluoroquinolone antibiotic and is an analog of norfloxacin. It targets gram-negative and gram-positive bacteria as well as several Mycoplasma species.

It is commonly used against Gonococcal species and to induce achilles tendon toxicity in rodents.

Chemical Identifiers

Molecular Weight (g/mol) 465.49 g/mol

Specifications

Boiling Point 529.1°C ±50.0°C
Quantity 10 g
Percent Purity >98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.