Learn More
Medchemexpress LLC PEG(2000)-C-DMG | 1443687-74-8 | 98.0% | 2600 g/mol (approx.) | 10 MG
SDP
Catalog No. 5000295146
Encompass_Preferred
PEG(2000)-C-DMG is a polyethylene glycol-conjugated lipid used as a PEG-lipid component in liposomes and lipid nanoparticles (LNPs). It helps stabilize particle surfaces, reduce protein adsorption, and modulate circulation time for nucleic acid delivery applications such as siRNA and mRNA formulations. Typical form is a white to off-white solid with solubility in DMSO (requires sonication).
  • Improves particle stability and circulation time.
  • Reduces serum protein adsorption and aggregation.
  • Suitable for lipid nanoparticle and liposome formulations for nucleic acid delivery.
  • Supplied as a solid that is soluble in DMSO with sonication.
  • High purity for research applications.
Catalog No. 50-002-95146 Supplier Medchemexpress LLC Supplier No. HY16839910MG
PEG(2000)-C-DMG is a polyethylene glycol-conjugated lipid used as a PEG-lipid component in liposomes and lipid nanoparticles (LNPs). It helps stabilize particle surfaces, reduce protein adsorption, and modulate circulation time for nucleic acid delivery applications such as siRNA and mRNA formulations. Typical form is a white to off-white solid with solubility in DMSO (requires sonication).
  • Improves particle stability and circulation time.
  • Reduces serum protein adsorption and aggregation.
  • Suitable for lipid nanoparticle and liposome formulations for nucleic acid delivery.
  • Supplied as a solid that is soluble in DMSO with sonication.
  • High purity for research applications.