Learn More
Cayman Chemical C-DMG-mPEG(2000), Purity: ≥95%, 25 mg
SDP
Catalog No. 502915156
Encompass_Preferred
A PEGylated derivative of DMG; has been used in combination with other lipids in the formation of LNPs for the delivery of siRNA; LNPs containing C-DMG-mPEG(2000) and encapsulating siRNA targeting the genes encoding PD-L1 and PD-L2 decrease PD-L1 and PD-L2 levels in monocyte-derived dendritic cells
Catalog No. 50-291-5156 Supplier Cayman Chemical Supplier No. 3714925 MG
A PEGylated derivative of DMG; has been used in combination with other lipids in the formation of LNPs for the delivery of siRNA; LNPs containing C-DMG-mPEG(2000) and encapsulating siRNA targeting the genes encoding PD-L1 and PD-L2 decrease PD-L1 and PD-L2 levels in monocyte-derived dendritic cells