Learn More
Cayman Chemical PEG 2000-dPE 10mg
SDP
Catalog No. 502915370
Encompass_Preferred
A PEGylated form of DSPE; has been used in the generation of micelles and in combination with other lipids in the formation of liposomes; liposomes containing DSPE-mPEG(2000) and encapsulating doxorubicin and quercetin reduce tumor growth in an MCF-7/adr multidrug-resistant breast cancer mouse xenograft model
Catalog No. 50-291-5370 Supplier Cayman Chemical Supplier No. 3714810 MG
A PEGylated form of DSPE; has been used in the generation of micelles and in combination with other lipids in the formation of liposomes; liposomes containing DSPE-mPEG(2000) and encapsulating doxorubicin and quercetin reduce tumor growth in an MCF-7/adr multidrug-resistant breast cancer mouse xenograft model