Learn More
Mitegen PEG 2000 MME, 50%(w/v), 100 ml, CAS# 9004-74-4
SDP
Catalog No. 501492083
Encompass_Preferred
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
Catalog No. 50-149-2083 Supplier Mitegen Supplier No. 1008066
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.