Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bioworld PEG 20%, 600 mL Liquid, 3.5 M Ammonium Acetate, Non-hazardous

Catalog No. NC2134316
Encompass_Preferred

This product is a precisely formulated buffer solution designed for various life science applications, particularly protein crystallization. It contains 20% polyethylene glycol (PEG) 8000 and 3.5 M ammonium acetate, offering consistent performance and a stable environment for experimental procedures. This ready-to-use liquid reagent simplifies preparation steps for researchers.

  • Contains 20% polyethylene glycol
  • Uses 3.5 M ammonium acetate
  • Engineered for protein crystallization
  • Ready-to-use liquid form
  • Stable composition ensures reproducibility
  • Non-hazardous for safe handling
  • Pack size of 600 mL (6 x 100 mL)
  • Has a shelf life of 2 years
  • Store at room temperature

Catalog No. NC2134316 Supplier Bioworld Supplier No. 401209361
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is a precisely formulated buffer solution designed for various life science applications, particularly protein crystallization. It contains 20% polyethylene glycol (PEG) 8000 and 3.5 M ammonium acetate, offering consistent performance and a stable environment for experimental procedures. This ready-to-use liquid reagent simplifies preparation steps for researchers.

  • Contains 20% polyethylene glycol
  • Uses 3.5 M ammonium acetate
  • Engineered for protein crystallization
  • Ready-to-use liquid form
  • Stable composition ensures reproducibility
  • Non-hazardous for safe handling
  • Pack size of 600 mL (6 x 100 mL)
  • Has a shelf life of 2 years
  • Store at room temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.