Learn More
Molecular Dimensions PEG 3350 250ML
SDP
Supplier:  Molecular Dimensions MD22509
Encompass
Protein Crystallography. 250mL bottle of 50% w/v PEG 3350 (Poly(ethylene glycol) 3350) solution. Made with ultra-purified water.
Catalog No. NC0620958