Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEG 35000, G-Biosciences
SCI_ED

Catalog No. 01001094
Change view
Click to view available options
Quantity:
5000 g, 1 mL
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-001-094 5000 g, 1 mL
01-001-084 250 g
01-001-086 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-001-094 Supplier G-Biosciences Supplier No. RC830
Add to Cart
Add to Cart

Polyethylene glycol (PEG) is a polyether compound that is used as an additive in cosmetics, pharmaceuticals, and food. It consists of repeated ethylene glycol units[-(CH2CH2O)n].

PEG has several biomedical applications including biosensing, imaging, and tissue engineering. It also repels protein adsorption and lowers the liver uptake, which helps in coating nanoparticles.

Chemical Identifiers

Molecular Weight (g/mol) 35,000 g/mol

Specifications

CAS 67-56-1, 1346746-84-6
Linear Formula C12H10D4BrNO2 · HCl
Quantity 5000 g, 1 mL
UN Number UN 1230 - class 3 - PG 2 - Methanol
Concentration 100 μg/mL in acetonitrile:water (9:1) (as free base)
Formula Weight 324.63
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder or Flakes
Product Line Gibco
Packaging Ampule
Flash Point 9.7°C (49.5°F) - closed cup
Show More Show Less
Hazard Category H311
Hazard Statement H331-H370
Recommended Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.