Learn More
LAB ALLEY Lab Alley Polyethylene Glycol (PEG) 400 Food Grade (FCC) Halal
SDP
Catalog No. NC2160118
Encompass
Polyethylene Glycol is synthesized by ring-opening polymerization of ethylene oxide. This synthesis process gives PEG a range of molecular weights and distributions from 300 grams per mole to 10000000 grams per mole. PEG 400 has a molecular weight of 400 and is a clear colorless viscous liquid. Polyethylene Glycol is colorless inert odorless and non-volatile. PEG is biocompatible (it will not damage tissues or cells) hydrophilic dissolves readily in water without changing the color odor or taste and is nontoxic. Due to the high purity of Polyethylene Glycol 400 it is certified by the United States Pharmacopeia (USP) grade and the Food Chemicals Codex (FCC) grade.
This item is not returnable. View return policy
Catalog No. NC2160118 Supplier LAB ALLEY Supplier No. PEG400F500ML
Polyethylene Glycol is synthesized by ring-opening polymerization of ethylene oxide. This synthesis process gives PEG a range of molecular weights and distributions from 300 grams per mole to 10000000 grams per mole. PEG 400 has a molecular weight of 400 and is a clear colorless viscous liquid. Polyethylene Glycol is colorless inert odorless and non-volatile. PEG is biocompatible (it will not damage tissues or cells) hydrophilic dissolves readily in water without changing the color odor or taste and is nontoxic. Due to the high purity of Polyethylene Glycol 400 it is certified by the United States Pharmacopeia (USP) grade and the Food Chemicals Codex (FCC) grade.