Learn More
Polysciences, Inc. PEG 4000 Resin, CAS No.: 25322-68-3, White Solid, This water soluble, nonionic material is used like paraffin to infiltrate and embed tissue
SDP
Catalog No. NC9756220
Encompass_Preferred
PEG 4000 Resin, CAS No.: 25322-68-3, White Solid, This water soluble, nonionic material is used like paraffin to infiltrate and embed tissue, Harmless-use normal precautions, Melting Point: 53 to 56 deg., Store at 25 deg.C
Catalog No. NC9756220 Supplier Polysciences, Inc. Supplier No. 16861250
PEG 4000 Resin, CAS No.: 25322-68-3, White Solid, This water soluble, nonionic material is used like paraffin to infiltrate and embed tissue, Harmless-use normal precautions, Melting Point: 53 to 56 deg., Store at 25 deg.C