Learn More
AG Scientific Inc PEG 8000, 1 KG
Catalog No. 502236861
Encompass_Preferred
PEG 8000 (Polyethylene Glycol) is ultra-pure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.CAS Number: 25322-68-3Molecular Weight: 7,000-9,000 DaSolubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.CAS Number: 25322-68-3Molecular Weight: 7,000-9,000 DaSolubility: WaterStorage Temperature: Room TemperatureShipping Weight: 0.0626 lbsResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6861 Supplier AG Scientific Inc Supplier No. P28291KG
PEG 8000 (Polyethylene Glycol) is ultra-pure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.CAS Number: 25322-68-3Molecular Weight: 7,000-9,000 DaSolubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.CAS Number: 25322-68-3Molecular Weight: 7,000-9,000 DaSolubility: WaterStorage Temperature: Room TemperatureShipping Weight: 0.0626 lbsResearch or further manufacturing use only, not for food or drug use.