Learn More
Research Products International Corp PEG 8000, 25 KG
SDP
Catalog No. 501977004
Encompass_Preferred
Ultrapure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.Melting Point: 55°-60°C
Catalog No. 50-197-7004 Supplier Research Products International Corp Supplier No. P4808025000.0
Ultrapure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.Melting Point: 55°-60°C