Learn More
Molecular Dimensions PEG 8000 250 MLS
SDP
Supplier:  Molecular Dimensions MD225013
Encompass
Protein Crystallography. 250mL bottle of 50% w/v PEG 8000 (Poly(ethylene glycol) 8000) solution. Made with ultra-purified water.
Catalog No. NC0365938