Learn More
Mitegen PEG 8000, 50%(w/v), 250 ml, CAS # 25322-68-3
SDP
Catalog No. NC9443505
Encompass_Preferred
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging.�Our stock solutions are intended for Research and Development Use Only.
This item is not returnable. View return policy
Catalog No. NC9443505 Supplier Mitegen Supplier No. 1008063
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging.�Our stock solutions are intended for Research and Development Use Only.