Learn More
Research Products International Corp PEG 8000 [Polyethylene Glycol 8000], 500 Grams
SDP
Catalog No. 50488949
Encompass_Preferred
Ultrapure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.Melting Point: 55°-60°C
Catalog No. 50-488-949 Supplier Research Products International Corp Supplier No. P48080500.0
Ultrapure white crystals for molecular biology applications such as precipitation of DNA, nucleic acids, hybridization and fusion of mammalian cells.Melting Point: 55°-60°C