Learn More
Alkali Scientific PEG 8000 [Polyethylene Glycol 8000], 10 Kg
SDP
Catalog No. 5000661232
Encompass_Preferred
Polyethylene Glycol 8000 (PEG 8000) is a high molecular weight polymer used extensively in biochemical and molecular biology applications. With a molecular weight of 8000, PEG 8000 is often employed to induce molecular crowding, a technique that simulates intracellular environments and promotes the formation of macromolecular complexes. It is ideal for protein precipitation, enzyme stabilization, and the enhancement of chemical reactions in vitro. This reagent is also used in DNA extraction protocols and protein solubilization studies. This product is offered in a 10 kg package, providing researchers with a large supply for repeated experiments and large-scale applications.
Catalog No. 50-006-61232 Supplier Alkali Scientific Supplier No. PEA002CP
Polyethylene Glycol 8000 (PEG 8000) is a high molecular weight polymer used extensively in biochemical and molecular biology applications. With a molecular weight of 8000, PEG 8000 is often employed to induce molecular crowding, a technique that simulates intracellular environments and promotes the formation of macromolecular complexes. It is ideal for protein precipitation, enzyme stabilization, and the enhancement of chemical reactions in vitro. This reagent is also used in DNA extraction protocols and protein solubilization studies. This product is offered in a 10 kg package, providing researchers with a large supply for repeated experiments and large-scale applications.