Learn More
Alkali Scientific PEG 8000 [Polyethylene Glycol 8000], 15 Kg
SDP
Catalog No. 5000661609
Encompass_Preferred
Polyethylene Glycol 8000 (PEG 8000) is a polymer used in laboratory applications to simulate molecular crowding environments. Its high molecular weight makes it useful for a variety of applications, such as enzyme stabilization, protein precipitation, and improving the solubility of macromolecules. PEG 8000 is also utilized in DNA extraction procedures and the preparation of high-concentration protein solutions. This reagent plays a key role in mimicking cellular environments, aiding in protein-protein interactions, and enhancing the efficiency of biochemical reactions. Available in a 15 kg package, it provides ample material for large-scale or extended research projects requiring high molecular weight PEG.
Catalog No. 50-006-61609 Supplier Alkali Scientific Supplier No. PEA003CP
Polyethylene Glycol 8000 (PEG 8000) is a polymer used in laboratory applications to simulate molecular crowding environments. Its high molecular weight makes it useful for a variety of applications, such as enzyme stabilization, protein precipitation, and improving the solubility of macromolecules. PEG 8000 is also utilized in DNA extraction procedures and the preparation of high-concentration protein solutions. This reagent plays a key role in mimicking cellular environments, aiding in protein-protein interactions, and enhancing the efficiency of biochemical reactions. Available in a 15 kg package, it provides ample material for large-scale or extended research projects requiring high molecular weight PEG.