Learn More
Alkali Scientific PEG 8000 [Polyethylene Glycol 8000], 25 Kg
SDP
Catalog No. 5000661610
Encompass_Preferred
Polyethylene Glycol 8000 (PEG 8000) is a high-molecular-weight polymer with a wide range of applications in molecular biology and biochemistry. This reagent, with a molecular weight of 8000, is primarily used for inducing molecular crowding, which helps study protein interactions and enhance enzyme stability. It is commonly used for protein precipitation, solubilizing membrane proteins, and in DNA extraction procedures. PEG 8000 is also crucial for macromolecular studies, such as DNA-protein interactions, and is often used in crystallography. Available in a 25 kg package, this product is designed to support large-scale research needs and repetitive experimental protocols.
Catalog No. 50-006-61610 Supplier Alkali Scientific Supplier No. PEA004CP
Polyethylene Glycol 8000 (PEG 8000) is a high-molecular-weight polymer with a wide range of applications in molecular biology and biochemistry. This reagent, with a molecular weight of 8000, is primarily used for inducing molecular crowding, which helps study protein interactions and enhance enzyme stability. It is commonly used for protein precipitation, solubilizing membrane proteins, and in DNA extraction procedures. PEG 8000 is also crucial for macromolecular studies, such as DNA-protein interactions, and is often used in crystallography. Available in a 25 kg package, this product is designed to support large-scale research needs and repetitive experimental protocols.