Learn More
Alkali Scientific PEG 8000 [Polyethylene Glycol 8000], 500 G
SDP
Catalog No. 5000661233
Encompass_Preferred
Polyethylene Glycol 8000 (PEG 8000) is a high-molecular-weight polymer frequently used in biochemical and molecular biology applications, especially for molecular crowding studies and enzyme stabilization. Its high molecular weight enhances protein solubility and promotes macromolecular interactions by mimicking the crowded environment of cells. PEG 8000 is also an important reagent for DNA extraction, protein precipitation, and other solubility-related applications. This product is available in a 500 g package, offering an ideal amount for small-scale research, enzyme reactions, and other biochemical assays that require a molecular crowding agent.
Catalog No. 50-006-61233 Supplier Alkali Scientific Supplier No. PEA005CP
Polyethylene Glycol 8000 (PEG 8000) is a high-molecular-weight polymer frequently used in biochemical and molecular biology applications, especially for molecular crowding studies and enzyme stabilization. Its high molecular weight enhances protein solubility and promotes macromolecular interactions by mimicking the crowded environment of cells. PEG 8000 is also an important reagent for DNA extraction, protein precipitation, and other solubility-related applications. This product is available in a 500 g package, offering an ideal amount for small-scale research, enzyme reactions, and other biochemical assays that require a molecular crowding agent.