Learn More
Medchemexpress LLC Peg-bis-amine | 24991-53-5 | 99.4% | (C2H4O)nC4H12N2O | 500 MG
SDP
Catalog No. 5000354935
Encompass_Preferred
PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs.
  • PROTACs are compounds containing two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets the protein of interest.
  • PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
Catalog No. 50-003-54935 Supplier Medchemexpress LLC Supplier No. HY140646500MG
PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker that can be utilized in the synthesis of PROTACs.
  • PROTACs are compounds containing two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets the protein of interest.
  • PROTACs leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.