Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PEG-bis-amine (MW 2000) | 24991-53-5 | 99.4% | (C2H4O)nC4H12N2O | 100 MG
SDP

Catalog No. 5000356450
Encompass_Preferred

PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only.

  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 2000 (average)

Catalog No. 50-003-56450 Supplier Medchemexpress LLC Supplier No. HY140646100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only.

  • Appearance: solid
  • Color: white to off-white
  • Molecular weight: 2000 (average)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.