Learn More
Medchemexpress LLC Peg-bis-amine (MW 2000) | 24991-53-5 | 99.4% | (C2H4O)nC4H12N2O | 250 MG
SDP
Catalog No. 5000352864
Encompass_Preferred
PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligands are for an E3 ubiquitin ligase and a target protein
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52864 Supplier Medchemexpress LLC Supplier No. 250MGHY140646
PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Can be used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligands are for an E3 ubiquitin ligase and a target protein
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins