Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PEG-bis-amine (MW 20000) | 24991-53-5 | 98.0% | (C2H4O)nC4H12N2O | 250 MG
SDP

Catalog No. 5000352774
Encompass_Preferred

PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 20000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein. This compound appears as a white to off-white solid and has an average molecular weight of 20000. It is intended for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • White to off-white solid appearance
  • Average molecular weight of 20000
  • Purity of 98.0%
  • Soluble in water at 100 mg/mL (requires ultrasonic)
  • Stable as powder at -20°C for 3 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-52774 Supplier Medchemexpress LLC Supplier No. 250MGHY140649
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 20000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules employ the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein. This compound appears as a white to off-white solid and has an average molecular weight of 20000. It is intended for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • White to off-white solid appearance
  • Average molecular weight of 20000
  • Purity of 98.0%
  • Soluble in water at 100 mg/mL (requires ultrasonic)
  • Stable as powder at -20°C for 3 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.