Learn More
Medchemexpress LLC PEG-bis-amine (MW 20000) | 24991-53-5 | 98.0% | (C2H4O)nC4H12N2O | 50 MG
SDP
Catalog No. 5000353353
Encompass_Preferred
PEG-bis-amine (MW 20000) is a poly(ethylene glycol)-based PROTAC linker, primarily utilized in the synthesis of Proteolysis Targeting Chimeras (PROTACs). These PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. The compound has an average molecular weight of 20000 and typically appears as a white to off-white solid, with a purity of 98.0%.
  • PEG-based linker for PROTAC synthesis
  • Enables targeted protein degradation via ubiquitin-proteasome system
  • White to off-white solid appearance
  • Average molecular weight of 20000
  • 98.0% purity
Catalog No. 50-003-53353 Supplier Medchemexpress LLC Supplier No. HY14064950MG
PEG-bis-amine (MW 20000) is a poly(ethylene glycol)-based PROTAC linker, primarily utilized in the synthesis of Proteolysis Targeting Chimeras (PROTACs). These PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins. The compound has an average molecular weight of 20000 and typically appears as a white to off-white solid, with a purity of 98.0%.
  • PEG-based linker for PROTAC synthesis
  • Enables targeted protein degradation via ubiquitin-proteasome system
  • White to off-white solid appearance
  • Average molecular weight of 20000
  • 98.0% purity