Learn More
Medchemexpress LLC PEG-bis-amine MW 80 1g
SDP
Catalog No. 5000206409
Encompass_Preferred
PEG-bis-amine (MW 8000) synthesizes folate-conjugated polymeric micelles for encapsulation of the anticancer agent 9-nitrocamptothecin HY-16560 (HY-16560) Folic acid-conjugated polymer micelles are effective carriers of insoluble anticancer drugs which can avoid macrophages and play a role in endocytosis of tumor cells mediated by folate receptors (FR)
Catalog No. 50-002-06409 Supplier Medchemexpress LLC Supplier No. HYW1156071G
PEG-bis-amine (MW 8000) synthesizes folate-conjugated polymeric micelles for encapsulation of the anticancer agent 9-nitrocamptothecin HY-16560 (HY-16560) Folic acid-conjugated polymer micelles are effective carriers of insoluble anticancer drugs which can avoid macrophages and play a role in endocytosis of tumor cells mediated by folate receptors (FR)