Learn More
Medchemexpress LLC PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) | 24991-53-5 | (C2H4O)nC4H12N2O | 1 G
SDP
Catalog No. 5000356451
Encompass_Preferred
PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-56451 Supplier Medchemexpress LLC Supplier No. HY1406461G
PEG-bis-amine (Poly(ethylene glycol)-bis-amine) (MW 2000) is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only