Learn More
BEZWADA BIOMEDICAL LLC PEG Diacrylate, MW 4000, 5.0 GM
SDP
Supplier:  BEZWADA BIOMEDICAL LLC 74K002
Encompass_Preferred
Acrylate/methacrylate end functionalization enables crosslinking. Polymers used in drug delivery and various biomedical applications
Catalog No. 50-190-4508