Learn More
Polysciences, Inc. Polyethylene glycol dimethacrylate (PEGDMA 400)
SDP
Catalog No. NC1929725
Encompass_Preferred
PEG-based cross-linked polymeric materials (hydrogels) are suitable carriers for drug delivery and various other biomedical applications.PEG 400 dimethacrylate (PEG400DMA) is a long-chain hydrophilic, crosslinking monomer. Molecular weight of PEG unit is approximately 400 (n=~9).
Catalog No. NC1929725 Supplier Polysciences, Inc. Supplier No. 151791
PEG-based cross-linked polymeric materials (hydrogels) are suitable carriers for drug delivery and various other biomedical applications.PEG 400 dimethacrylate (PEG400DMA) is a long-chain hydrophilic, crosslinking monomer. Molecular weight of PEG unit is approximately 400 (n=~9).