Learn More
Hampton Research PEG/ION 2 SCREEN #47A
SDP
Catalog No. 50563272
Encompass
HR2-998-47A/PEG/Ion 2 Screen #47A (H11), 185 ml - 1% w/v Tryptone, 0.05 M HEPES sodium pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-563-272 Supplier Hampton Research Supplier No. HR299847A
HR2-998-47A/PEG/Ion 2 Screen #47A (H11), 185 ml - 1% w/v Tryptone, 0.05 M HEPES sodium pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.