Learn More
Hampton Research HR2-998-01/PEG/Ion 2 Screen #01 (E01), 185 ml - 0.1 M Sodium malonate pH 4.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502453
Encompass
HR2-998-01/PEG/Ion 2 Screen #01 (E01), 185 ml - 0.1 M Sodium malonate pH 4.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-245-3 Supplier Hampton Research Supplier No. HR299801
HR2-998-01/PEG/Ion 2 Screen #01 (E01), 185 ml - 0.1 M Sodium malonate pH 4.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.