Learn More
Hampton Research HR2-998-02/PEG/Ion 2 Screen #02 (E02), 185 ml - 0.2 M Sodium malonate pH 4.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502454
Encompass
HR2-998-02/PEG/Ion 2 Screen #02 (E02), 185 ml - 0.2 M Sodium malonate pH 4.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-245-4 Supplier Hampton Research Supplier No. HR299802
HR2-998-02/PEG/Ion 2 Screen #02 (E02), 185 ml - 0.2 M Sodium malonate pH 4.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.