Learn More
Hampton Research HR2-998-03/PEG/Ion 2 Screen #03 (E03), 185 ml - 0.1 M Sodium malonate pH 5.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502455
Encompass
HR2-998-03/PEG/Ion 2 Screen #03 (E03), 185 ml - 0.1 M Sodium malonate pH 5.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-245-5 Supplier Hampton Research Supplier No. HR299803
HR2-998-03/PEG/Ion 2 Screen #03 (E03), 185 ml - 0.1 M Sodium malonate pH 5.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.