Learn More
Hampton Research HR2-998-04/PEG/Ion 2 Screen #04 (E04), 185 ml - 0.2 M Sodium malonate pH 5.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502456
Encompass
HR2-998-04/PEG/Ion 2 Screen #04 (E04), 185 ml - 0.2 M Sodium malonate pH 5.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-245-6 Supplier Hampton Research Supplier No. HR299804
HR2-998-04/PEG/Ion 2 Screen #04 (E04), 185 ml - 0.2 M Sodium malonate pH 5.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.