Learn More
Hampton Research HR2-998-11/PEG/Ion 2 Screen #11 (E11), 185 ml - 4% v/v Tacsimate pH 5.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502463
Encompass
HR2-998-11/PEG/Ion 2 Screen #11 (E11), 185 ml - 4% v/v Tacsimate pH 5.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-246-3 Supplier Hampton Research Supplier No. HR299811
Add to Cart
Add to Cart
HR2-998-11/PEG/Ion 2 Screen #11 (E11), 185 ml - 4% v/v Tacsimate pH 5.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.