Learn More
Hampton Research HR2-998-14/PEG/Ion 2 Screen #14 (F02), 185 ml - 8% v/v Tacsimate pH 6.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502466
Encompass
HR2-998-14/PEG/Ion 2 Screen #14 (F02), 185 ml - 8% v/v Tacsimate pH 6.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-246-6 Supplier Hampton Research Supplier No. HR299814
HR2-998-14/PEG/Ion 2 Screen #14 (F02), 185 ml - 8% v/v Tacsimate pH 6.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.