Learn More
Hampton Research HR2-998-15/PEG/Ion 2 Screen #15 (F03), 185 ml - 4% v/v Tacsimate pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502467
Encompass
HR2-998-15/PEG/Ion 2 Screen #15 (F03), 185 ml - 4% v/v Tacsimate pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-246-7 Supplier Hampton Research Supplier No. HR299815
HR2-998-15/PEG/Ion 2 Screen #15 (F03), 185 ml - 4% v/v Tacsimate pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.