Learn More
Hampton Research HR2-998-18/PEG/Ion 2 Screen #18 (F06), 185 ml - 8% v/v Tacsimate pH 8.0, 20% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502470
Encompass
HR2-998-18/PEG/Ion 2 Screen #18 (F06), 185 ml - 8% v/v Tacsimate pH 8.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-0 Supplier Hampton Research Supplier No. HR299818
HR2-998-18/PEG/Ion 2 Screen #18 (F06), 185 ml - 8% v/v Tacsimate pH 8.0, 20% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.