Learn More
Hampton Research HR2-998-19/PEG/Ion 2 Screen #19 (F07), 185 ml - 0.1 M Succinic acid pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502471
Encompass
HR2-998-19/PEG/Ion 2 Screen #19 (F07), 185 ml - 0.1 M Succinic acid pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-1 Supplier Hampton Research Supplier No. HR299819
HR2-998-19/PEG/Ion 2 Screen #19 (F07), 185 ml - 0.1 M Succinic acid pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.