Learn More
Hampton Research HR2-998-21/PEG/Ion 2 Screen #21 (F09), 185 ml - 0.1 M Ammonium citrate tribasic pH 7.0, 12% w/v Polyethylene glycol 3,350
SDP
Catalog No. 502473
Encompass
HR2-998-21/PEG/Ion 2 Screen #21 (F09), 185 ml - 0.1 M Ammonium citrate tribasic pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.
Catalog No. 50-247-3 Supplier Hampton Research Supplier No. HR299821
HR2-998-21/PEG/Ion 2 Screen #21 (F09), 185 ml - 0.1 M Ammonium citrate tribasic pH 7.0, 12% w/v Polyethylene glycol 3,350. Please note items from this supplier are non-returnable.